Product Details

SNP ID
rs34418207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31865727 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCAATATACTCCAAGATGACCCG[A/G]GCTATCTGCACAAGGGTCAGGATGA
Phenotype
MIM: 608272 MIM: 606107
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NEU1 PubMed Links

Gene Details

Gene
NEU1
Gene Name
neuraminidase 1 (lysosomal sialidase)
There are no transcripts associated with this gene.

Gene
SLC44A4
Gene Name
solute carrier family 44 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178044.1 1614 Silent Mutation GCC,GCT A473A NP_001171515.1
NM_001178045.1 1614 Silent Mutation GCC,GCT A439A NP_001171516.1
NM_025257.2 1614 Silent Mutation GCC,GCT A515A NP_079533.2

View Full Product Details