Product Details

SNP ID
rs4802567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49129243 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGGCAGATTGGATAGCTGGTTG[C/T]GGCTGCATACCCGTTATGGAGACAG
Phenotype
MIM: 603144
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPFIA3 PubMed Links

Gene Details

Gene
PPFIA3
Gene Name
PTPRF interacting protein alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003660.3 Intron NP_003651.1
XM_017027407.1 Intron XP_016882896.1

View Full Product Details