Product Details

SNP ID
rs751077492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:175074912 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGTATGCTGAGGCAATACACAG[A/T]GTAAAAAGTTAGAAAGTATCTTACA
Phenotype
MIM: 123811
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ATF2 PubMed Links

Gene Details

Gene
ATF2
Gene Name
activating transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256090.1 Intron NP_001243019.1
NM_001256091.1 Intron NP_001243020.1
NM_001256092.1 Intron NP_001243021.1
NM_001256093.1 Intron NP_001243022.1
NM_001256094.1 Intron NP_001243023.1
NM_001880.3 Intron NP_001871.2

View Full Product Details