Product Details

SNP ID
rs6474226
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:40153560 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAGCAAAGCGAAGCCACCAAGCC[A/G]TCATCATGTCCACGTCGCTACGAGT
Phenotype
MIM: 607702
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C8orf4 PubMed Links

Gene Details

Gene
C8orf4
Gene Name
chromosome 8 open reading frame 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020130.4 93 Missense Mutation ATC,GTC I10V NP_064515.1

View Full Product Details