Product Details

SNP ID
rs12621643
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:223053265 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATTCAGGAGGAGGGGGCAGACGA[G/T]GAGCTGGAGGAGACCTCGGAGACGC
Phenotype
MIM: 607775
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KCNE4 PubMed Links

Gene Details

Gene
KCNE4
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080671.3 803 Missense Mutation GAG,GAT E196D NP_542402.3

View Full Product Details