Product Details

SNP ID
rs116034004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:104348588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACCTCAGGCACTGATATCTTCCT[A/G]AGTCTTTGGGAGATTTACGTGTCTC
Phenotype
MIM: 605933
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCSTAMP PubMed Links

Gene Details

Gene
DCSTAMP
Gene Name
dendrocyte expressed seven transmembrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257317.1 115 Silent Mutation CTA,CTG L12L NP_001244246.1
NM_030788.3 115 Silent Mutation CTA,CTG L12L NP_110415.1
XM_011517321.1 115 Silent Mutation CTA,CTG L12L XP_011515623.1
XM_011517324.1 115 Silent Mutation CTA,CTG L12L XP_011515626.1
XM_017013878.1 115 Silent Mutation CTA,CTG L12L XP_016869367.1
XM_017013879.1 115 Silent Mutation CTA,CTG L12L XP_016869368.1
XM_017013880.1 115 Silent Mutation CTA,CTG L12L XP_016869369.1

View Full Product Details