Product Details

SNP ID
rs45575940
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:46468516 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AGAGAGAACTTGGTTGACTTGGTAC[G/T]TCTTCCTTGTTCTTTCAACTCCTCA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PNMAL1 PubMed Links

Gene Details

Gene
PNMAL1
Gene Name
paraneoplastic Ma antigen family like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001103149.1 1756 UTR 3 NP_001096619.1
NM_018215.3 1756 UTR 3 NP_060685.2
XM_011527067.2 1756 UTR 3 XP_011525369.1

View Full Product Details