Product Details

SNP ID
rs61737320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:37990858 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACATCATGTCAGAGAAAAAGAAC[G/T]GTAAGAACTCGTCTACAAATAACAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HYPM PubMed Links

Gene Details

Gene
HYPM
Gene Name
huntingtin interacting protein M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012274.1 42 Missense Mutation GGT,TGT G7C NP_036406.1
Gene
SYTL5
Gene Name
synaptotagmin like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163334.1 42 Intron NP_001156806.1
NM_001163335.1 42 Intron NP_001156807.1
NM_138780.2 42 Intron NP_620135.1
XM_011544001.2 42 Intron XP_011542303.1
XM_011544002.2 42 Intron XP_011542304.1
XM_017029972.1 42 Intron XP_016885461.1
XM_017029973.1 42 Intron XP_016885462.1
XM_017029974.1 42 Intron XP_016885463.1

View Full Product Details