Product Details

SNP ID
rs77336931
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:206960399 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGCAGATGTGGGGCCCCCCAAC[C/T]GGGAGAAGGTGCCTGGGGTTACCGC
Phenotype
MIM: 605484
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FCAMR PubMed Links

Gene Details

Gene
FCAMR
Gene Name
Fc fragment of IgA and IgM receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122979.2 Intron NP_001116451.1
NM_001170631.1 Intron NP_001164102.1
NM_032029.4 Intron NP_114418.2
XM_011510051.2 Intron XP_011508353.1
XM_011510052.2 Intron XP_011508354.1
XM_017002442.1 Intron XP_016857931.1

View Full Product Details