Product Details

SNP ID
rs34478566
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:206960429 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGTGCCTGGGGTTACCGCTTCA[C/T]GGAGGACTCCTTGCCAGGGGGTCCC
Phenotype
MIM: 605484
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FCAMR PubMed Links

Gene Details

Gene
FCAMR
Gene Name
Fc fragment of IgA and IgM receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122979.2 1657 Intron NP_001116451.1
NM_001170631.1 1657 Missense Mutation ATG,GTG M483V NP_001164102.1
NM_032029.4 1657 Intron NP_114418.2
XM_011510051.2 1657 Missense Mutation ATG,GTG M483V XP_011508353.1
XM_011510052.2 1657 Missense Mutation ATG,GTG M471V XP_011508354.1
XM_017002442.1 1657 Missense Mutation ATG,GTG M483V XP_016857931.1

View Full Product Details