Product Details

SNP ID
rs1024940040
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57451298 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTCTCTCCTCTCCATCCCCTTT[G/T]GTACCTTCCTTCATTTCCAAGGCCA
Phenotype
MIM: 608943 MIM: 612837
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CIAPIN1 PubMed Links

Gene Details

Gene
CIAPIN1
Gene Name
cytokine induced apoptosis inhibitor 1
There are no transcripts associated with this gene.

Gene
COQ9
Gene Name
coenzyme Q9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020312.3 Intron NP_064708.1

View Full Product Details