Product Details

SNP ID
hCV25644987
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:139414128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAACAAGTAGCAAAACCCCAGCCC[G/T]GCGTAGGACCATTATCCTCTCAGCC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC18 PubMed Links

Gene Details

Gene
DNAJC18
Gene Name
DnaJ heat shock protein family (Hsp40) member C18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152686.3 1177 UTR 3 NP_689899.1
Gene
SPATA24
Gene Name
spermatogenesis associated 24
There are no transcripts associated with this gene.

View Full Product Details