Product Details

SNP ID
rs8177907
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:81087641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTCCCAACTTCAGGGAGACTACA[A/G]TAAACAACATCTCCCAACGCTTCCT
Phenotype
MIM: 238330
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GCSH PubMed Links

Gene Details

Gene
GCSH
Gene Name
glycine cleavage system protein H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004483.4 349 Silent Mutation TAC,TAT Y84Y NP_004474.2
XM_017023136.1 349 Intron XP_016878625.1
XM_017023137.1 349 Intron XP_016878626.1

View Full Product Details