Product Details

SNP ID
rs34514408
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:156244850 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCAGCGCGCAGAAGAGATGGTAG[A/C]CATGGCTGGTGCTCAGGGCCTCCTG
Phenotype
MIM: 112260 MIM: 614579 MIM: 609176 MIM: 610962
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BGLAP PubMed Links

Gene Details

Gene
BGLAP
Gene Name
bone gamma-carboxyglutamate protein
There are no transcripts associated with this gene.

Gene
PAQR6
Gene Name
progestin and adipoQ receptor family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272104.1 899 Missense Mutation NP_001259033.1
NM_001272105.1 899 Missense Mutation NP_001259034.1
NM_001272106.1 899 Intron NP_001259035.1
NM_001272107.1 899 Missense Mutation NP_001259036.1
NM_001272108.1 899 Missense Mutation NP_001259037.1
NM_001272109.1 899 Missense Mutation NP_001259038.1
NM_001272110.1 899 Missense Mutation NP_001259039.1
NM_001272111.1 899 Missense Mutation NP_001259040.1
NM_001272112.1 899 Missense Mutation NP_001259041.1
NM_001272113.1 899 Missense Mutation NP_001259042.1
NM_024897.3 899 Missense Mutation NP_079173.2
NM_198406.2 899 Missense Mutation NP_940798.1
XM_005245494.3 899 Missense Mutation XP_005245551.1
XM_006711546.2 899 Missense Mutation XP_006711609.1
XM_006711547.3 899 Missense Mutation XP_006711610.1
XM_006711548.3 899 Missense Mutation XP_006711611.1
XM_006711552.2 899 Missense Mutation XP_006711615.1
XM_006711553.3 899 Missense Mutation XP_006711616.1
XM_011510000.2 899 Missense Mutation XP_011508302.1
XM_017002385.1 899 Missense Mutation XP_016857874.1
Gene
PMF1
Gene Name
polyamine-modulated factor 1
There are no transcripts associated with this gene.

Gene
PMF1-BGLAP
Gene Name
PMF1-BGLAP readthrough
There are no transcripts associated with this gene.

Gene
SMG5
Gene Name
SMG5, nonsense mediated mRNA decay factor
There are no transcripts associated with this gene.

View Full Product Details