Product Details

SNP ID
rs56997592
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20112864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGGGCTCCTGTGGGAGCTGGGG[A/G]CTTGGTAGCAGGCCTAGCCCCCACC
Phenotype
MIM: 609030 MIM: 601180 MIM: 611151
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DGCR8 PubMed Links

Gene Details

Gene
DGCR8
Gene Name
DGCR8 microprocessor complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190326.1 Intron NP_001177255.1
NM_022720.6 Intron NP_073557.3
XM_006724268.2 Intron XP_006724331.1
Gene
MIR6816
Gene Name
microRNA 6816
There are no transcripts associated with this gene.

Gene
RANBP1
Gene Name
RAN binding protein 1
There are no transcripts associated with this gene.

Gene
TRMT2A
Gene Name
tRNA methyltransferase 2 homolog A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257994.1 Intron NP_001244923.1
NM_022727.5 Intron NP_073564.3
NM_182984.4 Intron NP_892029.2
XM_011530139.2 Intron XP_011528441.1
XM_011530141.1 Intron XP_011528443.1
XM_011530142.2 Intron XP_011528444.1
XM_017028770.1 Intron XP_016884259.1

View Full Product Details