Product Details
- SNP ID
-
rs61731283
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:57808466 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCACATATCCCACACGTATAAGG[C/T]TTTTCTCTAGTGTGAACTCCTTGAT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF552
PubMed Links
Gene Details
- Gene
- ZNF552
- Gene Name
- zinc finger protein 552
View Full Product Details