Product Details
- SNP ID
-
hCV25649859
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:79594496 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTGTTTATTTATAATCTTCCTAAG[G/T]GGCATATATCTCCCCAATTTGGTAA
- Phenotype
-
MIM: 601936
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PAWR
PubMed Links
Gene Details
- Gene
- PAWR
- Gene Name
- pro-apoptotic WT1 regulator
There are no transcripts associated with this gene.
View Full Product Details