Product Details

SNP ID
rs202205648
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:57974911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGTTAGCAGCTTCTAAAATAAG[A/T]AGAAAAAGGGTCTTTAGATTCTCAA
Phenotype
MIM: 606747
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
VEZF1 PubMed Links

Gene Details

Gene
VEZF1
Gene Name
vascular endothelial zinc finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007146.2 Intron NP_009077.2
XM_005257643.1 Intron XP_005257700.1
XM_005257644.4 Intron XP_005257701.1
XM_017025017.1 Intron XP_016880506.1
XM_017025018.1 Intron XP_016880507.1

View Full Product Details