Product Details

SNP ID
rs34536451
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:102439016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTCCTGCCTGCCCTGAGCCCGG[C/T]CATAAGGGCATGGATTACCGGGACT
Phenotype
MIM: 610929
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ORAI2 PubMed Links

Gene Details

Gene
ORAI2
Gene Name
ORAI calcium release-activated calcium modulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126340.2 205 Silent Mutation GGC,GGT G20G NP_001119812.1
NM_001271818.1 205 Silent Mutation GGC,GGT G20G NP_001258747.1
NM_001271819.1 205 Intron NP_001258748.1
NM_032831.3 205 Silent Mutation GGC,GGT G20G NP_116220.1

View Full Product Details