Product Details

SNP ID
rs17847933
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33198003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGACACCTTCACCATTTAGTCT[C/G]TTTCCAATCTCCCCCTAGCAAAACT
Phenotype
MIM: 120290 MIM: 601417 MIM: 611500 MIM: 180246 MIM: 601416
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
COL11A2 PubMed Links

Gene Details

Gene
COL11A2
Gene Name
collagen type XI alpha 2
There are no transcripts associated with this gene.

Gene
HSD17B8
Gene Name
hydroxysteroid 17-beta dehydrogenase 8
There are no transcripts associated with this gene.

Gene
MIR219A1
Gene Name
microRNA 219a-1
There are no transcripts associated with this gene.

Gene
RXRB
Gene Name
retinoid X receptor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270401.1 Intron NP_001257330.1
NM_001291989.1 Intron NP_001278918.1
NM_021976.4 Intron NP_068811.1
XM_005249278.2 Intron XP_005249335.1
XM_011514796.2 Intron XP_011513098.1
XM_017011176.1 Intron XP_016866665.1
Gene
SLC39A7
Gene Name
solute carrier family 39 member 7
There are no transcripts associated with this gene.

View Full Product Details