Product Details

SNP ID
rs948332215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:106803130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTCTTCTCCCGACACCACCTTTC[C/T]GCCACCACCTCCAAGTCCTGAGAAT
Phenotype
MIM: 300439
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF128 PubMed Links

Gene Details

Gene
RNF128
Gene Name
ring finger protein 128, E3 ubiquitin protein ligase
There are no transcripts associated with this gene.

Gene
TBC1D8B
Gene Name
TBC1 domain family member 8B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017752.2 Intron NP_060222.2
NM_198881.1 Intron NP_942582.1

View Full Product Details