Product Details

SNP ID
rs61734646
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128123014 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACGGCACCCTCCACGGCTCCGAA[C/G]TTGCCCTCGCGGACAATGTAGTCAA
Phenotype
MIM: 608152
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PTGES2 PubMed Links

Gene Details

Gene
PTGES2
Gene Name
prostaglandin E synthase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256335.1 836 Missense Mutation AAC,AAG N78K NP_001243264.1
NM_025072.6 836 Missense Mutation AAC,AAG N269K NP_079348.1
NM_198938.2 836 Missense Mutation AAC,AAG N78K NP_945176.1
XM_005252239.2 836 Missense Mutation AAC,AAG N78K XP_005252296.1
XM_011519050.1 836 Missense Mutation AAC,AAG N78K XP_011517352.1
XM_011519051.2 836 Missense Mutation AAC,AAG N78K XP_011517353.1
Gene
PTGES2-AS1
Gene Name
PTGES2 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
SLC25A25-AS1
Gene Name
SLC25A25 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details