Product Details

SNP ID
rs2466620
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124319410 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTTTTTCCATAGTAGTGCTGGAA[A/G]GGGGCTTGAAATACATGAATGTTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR8D1 PubMed Links

Gene Details

Gene
OR8D1
Gene Name
olfactory receptor family 8 subfamily D member 1
There are no transcripts associated with this gene.

Gene
OR8D2
Gene Name
olfactory receptor family 8 subfamily D member 2 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002918.1 788 Missense Mutation CCT,CTT P263L NP_001002918.1
Gene
OR8G5
Gene Name
olfactory receptor family 8 subfamily G member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005198.1 788 Intron NP_001005198.1
XM_011542662.2 788 Intron XP_011540964.1
XM_011542663.2 788 Intron XP_011540965.1
XM_011542664.2 788 Intron XP_011540966.1
XM_011542665.2 788 Intron XP_011540967.1
XM_011542666.2 788 Intron XP_011540968.1
XM_011542667.2 788 Intron XP_011540969.1

View Full Product Details