Product Details

SNP ID
hCV25747763
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:145846514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGACCCATGCCTGGGACCATCTT[T/G]GGTGTGGGCTGAGGGAGAGGCAAGC
Phenotype
MIM: 605987
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR6736 PubMed Links

Gene Details

Gene
MIR6736
Gene Name
microRNA 6736
There are no transcripts associated with this gene.

Gene
NUDT17
Gene Name
nudix hydrolase 17
There are no transcripts associated with this gene.

Gene
PIAS3
Gene Name
protein inhibitor of activated STAT 3
There are no transcripts associated with this gene.

Gene
POLR3C
Gene Name
RNA polymerase III subunit C
There are no transcripts associated with this gene.

View Full Product Details