Product Details

SNP ID
hCV25749022
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:202138286 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCTCCTCCCAGGGGCCTCCGTTG[C/G]CCTCCCCAACACACACACACACACA
Phenotype
MIM: 616597 MIM: 176889
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARL8A PubMed Links

Gene Details

Gene
ARL8A
Gene Name
ADP ribosylation factor like GTPase 8A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256129.1 Intron NP_001243058.1
NM_138795.3 Intron NP_620150.1
Gene
GPR37L1
Gene Name
G protein-coupled receptor 37 like 1
There are no transcripts associated with this gene.

Gene
PTPN7
Gene Name
protein tyrosine phosphatase, non-receptor type 7
There are no transcripts associated with this gene.

View Full Product Details