Product Details

SNP ID
rs35012530
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:92615990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTCTCGTAGGGCTTGAGGTCAA[C/G]GTCCAGCACAGACACGGAAAAGGCA
Phenotype
MIM: 611505
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CENPP PubMed Links

Gene Details

Gene
CENPP
Gene Name
centromere protein P
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012267.2 1595 Intron NP_001012267.1
NM_001286969.1 1595 Intron NP_001273898.1
NM_001286971.1 1595 Intron NP_001273900.1
XM_011518685.2 1595 Intron XP_011516987.1
XM_011518689.1 1595 Intron XP_011516991.1
XM_017014715.1 1595 Intron XP_016870204.1
Gene
IPPK
Gene Name
inositol-pentakisphosphate 2-kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022755.5 1595 Missense Mutation CTT,GTT L440V NP_073592.1
XM_017015041.1 1595 UTR 3 XP_016870530.1
Gene
LOC100128361
Gene Name
uncharacterized LOC100128361
There are no transcripts associated with this gene.

View Full Product Details