Product Details

SNP ID
rs373229392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:41062622 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTAAGTTTTACATGAGCTTAG[C/T]CTTGTTGTCTCTCTTTTTCAGGGCG
Phenotype
MIM: 189973 MIM: 604981
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ELF1 PubMed Links

Gene Details

Gene
ELF1
Gene Name
E74 like ETS transcription factor 1
There are no transcripts associated with this gene.

Gene
WBP4
Gene Name
WW domain binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007187.3 Intron NP_009118.1
XM_005266245.2 Intron XP_005266302.1
XM_017020380.1 Intron XP_016875869.1

View Full Product Details