Product Details

SNP ID
rs28384478
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:119791487 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTTTAAAAGGCTCCCGCCCCTC[A/G]ACGCTCCGCGCGGCCCAGGCATTTC
Phenotype
MIM: 300899
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPL39 PubMed Links

Gene Details

Gene
RPL39
Gene Name
ribosomal protein L39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001000.3 Intron NP_000991.1
Gene
SNORA69
Gene Name
small nucleolar RNA, H/ACA box 69
There are no transcripts associated with this gene.

View Full Product Details