Product Details

SNP ID
rs34255722
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75932958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTCCTAGTGTTCCCTTATTCGG[C/T]TTCTTTGGAAATGGAGAAATTGGAT
Phenotype
MIM: 609096 MIM: 610894
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXO22 PubMed Links

Gene Details

Gene
FBXO22
Gene Name
F-box protein 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012170.3 1173 Intron NP_036302.1
NM_147188.2 1173 Silent Mutation GGC,GGT G356G NP_671717.1
XM_011521447.2 1173 Silent Mutation GGC,GGT G252G XP_011519749.1
Gene
FBXO22-AS1
Gene Name
FBXO22 antisense RNA 1
There are no transcripts associated with this gene.

Gene
NRG4
Gene Name
neuregulin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138573.3 1173 Intron NP_612640.1

View Full Product Details