Product Details

SNP ID
rs2290417
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143575099 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGGGAGCGGCTCACACAGCTGCA[C/G]GGAGGAGGTAAGGAAAGAGAAGCTT
Phenotype
MIM: 130592 MIM: 611552
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
EEF1D PubMed Links
Additional Information
For this assay, SNP(s) [rs151207487] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
EEF1D
Gene Name
eukaryotic translation elongation factor 1 delta
There are no transcripts associated with this gene.

Gene
MROH6
Gene Name
maestro heat like repeat family member 6
There are no transcripts associated with this gene.

Gene
NAPRT
Gene Name
nicotinate phosphoribosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286829.1 Intron NP_001273758.1
NM_145201.5 Intron NP_660202.3
XM_017013975.1 Intron XP_016869464.1
XM_017013976.1 Intron XP_016869465.1
XM_017013977.1 Intron XP_016869466.1
XM_017013978.1 Intron XP_016869467.1
XM_017013979.1 Intron XP_016869468.1
XM_017013980.1 Intron XP_016869469.1

View Full Product Details