Product Details

SNP ID
rs35064727
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:42903302 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTGAATAAGGGCAGGAAGGTAG[C/T]TAGTCCCAGTCCCAGTGGCAGCGTG
Phenotype
MIM: 602184
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDUFV3 PubMed Links

Gene Details

Gene
NDUFV3
Gene Name
NADH:ubiquinone oxidoreductase subunit V3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001503.1 356 Intron NP_001001503.1
NM_021075.3 356 Missense Mutation GCT,GTT A97V NP_066553.3
XM_011529586.2 356 Missense Mutation GCT,GTT A97V XP_011527888.1
XM_017028359.1 356 Intron XP_016883848.1

View Full Product Details