Product Details

SNP ID
rs1046653667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:12673295 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTTTAAAATAACAAACCTCTAA[A/T]TAGCTAAGTAATGTACAATGTGTAA
Phenotype
MIM: 609702
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CEP76 PubMed Links

Gene Details

Gene
CEP76
Gene Name
centrosomal protein 76
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271989.1 2279 UTR 3 NP_001258918.1
NM_024899.3 2279 UTR 3 NP_079175.2
Gene
PSMG2
Gene Name
proteasome assembly chaperone 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020232.4 2279 Intron NP_064617.2
NM_147163.1 2279 Intron NP_671692.1

View Full Product Details