Product Details

SNP ID
rs958326741
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:46032963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTGAGCTATTTGCCACTGCCAG[C/T]AGATGAGTATGATGAGCAACCAAGA
Phenotype
MIM: 612452 MIM: 157140
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KANSL1 PubMed Links

Gene Details

Gene
KANSL1
Gene Name
KAT8 regulatory NSL complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193465.1 Intron NP_001180394.1
NM_001193466.1 Intron NP_001180395.1
NM_015443.3 Intron NP_056258.1
Gene
MAPT
Gene Name
microtubule associated protein tau
There are no transcripts associated with this gene.

View Full Product Details