Product Details

SNP ID
rs149097782
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:41061810 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGCGTTCTTAACTCGCTCGGGAC[C/T]GGCCCCAGACCTGCAGGGCCGGTTC
Phenotype
MIM: 189973 MIM: 604981
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ELF1 PubMed Links

Gene Details

Gene
ELF1
Gene Name
E74 like ETS transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145353.1 Intron NP_001138825.1
NM_172373.3 Intron NP_758961.1
XM_005266276.1 Intron XP_005266333.1
XM_005266277.3 Intron XP_005266334.1
XM_011534950.1 Intron XP_011533252.1
XM_011534951.2 Intron XP_011533253.1
XM_017020409.1 Intron XP_016875898.1
XM_017020410.1 Intron XP_016875899.1
XM_017020411.1 Intron XP_016875900.1
XM_017020412.1 Intron XP_016875901.1
XM_017020413.1 Intron XP_016875902.1
Gene
WBP4
Gene Name
WW domain binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007187.3 Intron NP_009118.1
XM_005266245.2 Intron XP_005266302.1
XM_017020380.1 Intron XP_016875869.1

View Full Product Details