Product Details

SNP ID
rs114825631
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19526169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCGGGCTGGCCCGCTGAGCAGCG[C/T]CTCTCTTCACAGGCTACAGCATGCT
Phenotype
MIM: 609435
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDUFA13 PubMed Links

Gene Details

Gene
NDUFA13
Gene Name
NADH:ubiquinone oxidoreductase subunit A13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015965.6 Intron NP_057049.5
Gene
YJEFN3
Gene Name
YjeF N-terminal domain containing 3
There are no transcripts associated with this gene.

View Full Product Details