Product Details

SNP ID
rs3816244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:163506833 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTGGAGAGCACCTCTGAATATATC[A/G]TTTCTGTCATTACCATGGCAACAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMA16 PubMed Links

Gene Details

Gene
TMA16
Gene Name
translation machinery associated 16 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018352.2 Intron NP_060822.2

View Full Product Details