Product Details

SNP ID
hCV25756256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43827299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACATCTGGGATTCCCTTTCTTCA[A/C]CCTTCTGCGTTTTCTTCTGCCACAG
Phenotype
MIM: 600215
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MFAP1 PubMed Links

Gene Details

Gene
MFAP1
Gene Name
microfibrillar associated protein 1
There are no transcripts associated with this gene.

Gene
WDR76
Gene Name
WD repeat domain 76
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167941.1 Intron NP_001161413.1
NM_024908.3 Intron NP_079184.2

View Full Product Details