Product Details

SNP ID
rs35602166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241495488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCCTGACGTGCACAACAGCACAC[C/T]GCAGGGGTGGAAGGAGACGGTGACC
Phenotype
MIM: 602255
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FARP2 PubMed Links

Gene Details

Gene
FARP2
Gene Name
FERM, ARH/RhoGEF and pleckstrin domain protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282983.1 1514 Intron NP_001269912.1
NM_001282984.1 1514 Intron NP_001269913.1
NM_014808.3 1514 Intron NP_055623.1
XM_005247050.3 1514 Intron XP_005247107.1
XM_011512233.1 1514 Intron XP_011510535.1
XM_011512234.1 1514 Intron XP_011510536.1
XM_011512235.1 1514 Intron XP_011510537.1
XM_011512236.1 1514 Intron XP_011510538.1
XM_017005416.1 1514 Intron XP_016860905.1
XM_017005417.1 1514 Intron XP_016860906.1
XM_017005418.1 1514 Intron XP_016860907.1
XM_017005419.1 1514 Intron XP_016860908.1
XM_017005420.1 1514 Intron XP_016860909.1
Gene
STK25
Gene Name
serine/threonine kinase 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271977.1 1514 UTR 3 NP_001258906.1
NM_001271978.1 1514 UTR 3 NP_001258907.1
NM_001271979.1 1514 UTR 3 NP_001258908.1
NM_001271980.1 1514 UTR 3 NP_001258909.1
NM_001282305.1 1514 UTR 3 NP_001269234.1
NM_001282306.1 1514 UTR 3 NP_001269235.1
NM_001282307.1 1514 UTR 3 NP_001269236.1
NM_001282308.1 1514 UTR 3 NP_001269237.1
NM_006374.4 1514 UTR 3 NP_006365.2
XM_011510493.2 1514 Intron XP_011508795.1
XM_011510494.2 1514 UTR 3 XP_011508796.1
XM_011510495.2 1514 UTR 3 XP_011508797.1
XM_011510496.2 1514 Intron XP_011508798.1
XM_017003168.1 1514 UTR 3 XP_016858657.1
XM_017003169.1 1514 UTR 3 XP_016858658.1
XM_017003170.1 1514 UTR 3 XP_016858659.1
XM_017003171.1 1514 UTR 3 XP_016858660.1

View Full Product Details