Product Details

SNP ID
rs34799130
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60429700 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGATTCAAGCACCGCACACAGTC[C/T]GGTGTTTCTGGTATTTCCTCCAGAA
Phenotype
MIM: 606499
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MS4A5 PubMed Links

Gene Details

Gene
MS4A5
Gene Name
membrane spanning 4-domains A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023945.2 112 Missense Mutation CCG,CTG P9L NP_076434.2

View Full Product Details