Product Details
- SNP ID
-
rs35828562
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:1443792 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATGACAGCATTGAGCAGGTTGGCT[C/T]CCTTCAGGGTCTGGCTCTCATCAGC
- Phenotype
-
MIM: 606610
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NSFL1C
PubMed Links
Gene Details
- Gene
- NSFL1C
- Gene Name
- NSFL1 cofactor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001206736.1 |
2495 |
Missense Mutation |
GAA,GGA |
E359G |
NP_001193665.1 |
NM_016143.4 |
2495 |
Missense Mutation |
GAA,GGA |
E357G |
NP_057227.2 |
NM_018839.4 |
2495 |
Missense Mutation |
GAA,GGA |
E326G |
NP_061327.2 |
XM_006723592.2 |
2495 |
Missense Mutation |
GAA,GGA |
E248G |
XP_006723655.1 |
XM_006723593.2 |
2495 |
Missense Mutation |
GAA,GGA |
E248G |
XP_006723656.1 |
XM_006723594.2 |
2495 |
Missense Mutation |
GAA,GGA |
E246G |
XP_006723657.1 |
XM_011529300.1 |
2495 |
Missense Mutation |
GAA,GGA |
E362G |
XP_011527602.1 |
XM_011529301.1 |
2495 |
Missense Mutation |
GAA,GGA |
E360G |
XP_011527603.1 |
XM_011529302.2 |
2495 |
Missense Mutation |
GAA,GGA |
E309G |
XP_011527604.1 |
XM_011529304.1 |
2495 |
Missense Mutation |
GAA,GGA |
E306G |
XP_011527606.1 |
XM_011529305.2 |
2495 |
Intron |
|
|
XP_011527607.1 |
XM_017027956.1 |
2495 |
Missense Mutation |
GAA,GGA |
E307G |
XP_016883445.1 |
XM_017027957.1 |
2495 |
Missense Mutation |
GAA,GGA |
E308G |
XP_016883446.1 |
XM_017027958.1 |
2495 |
Missense Mutation |
GAA,GGA |
E272G |
XP_016883447.1 |
View Full Product Details