Product Details

SNP ID
rs61736664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:127524531 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTATCCTCTCCACTTTTTTCTAG[C/T]GGCAACCAGTTCTTTTTCTTCTCCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRRC1 PubMed Links

Gene Details

Gene
PRRC1
Gene Name
proline rich coiled-coil 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286808.1 292 Missense Mutation GCG,GTG A35V NP_001273737.1
NM_130809.4 292 Missense Mutation GCG,GTG A35V NP_570721.1
XM_006714529.3 292 Missense Mutation GCG,GTG A35V XP_006714592.1

View Full Product Details