Product Details

SNP ID
rs3817731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45890204 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCTACGGAGCCCTAGTAGATTCC[A/T]GTCCCCACACTGAGCCAGGAGGGGT
Phenotype
MIM: 613111 MIM: 608597 MIM: 172425
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CTSA PubMed Links

Gene Details

Gene
CTSA
Gene Name
cathepsin A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000308.3 61 UTR 5 NP_000299.2
NM_001127695.2 61 UTR 5 NP_001121167.1
NM_001167594.2 61 UTR 5 NP_001161066.1
Gene
NEURL2
Gene Name
neuralized E3 ubiquitin protein ligase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278535.1 61 Intron NP_001265464.1
NM_080749.3 61 Intron NP_542787.1
Gene
PLTP
Gene Name
phospholipid transfer protein
There are no transcripts associated with this gene.

Gene
SPATA25
Gene Name
spermatogenesis associated 25
There are no transcripts associated with this gene.

Gene
ZSWIM1
Gene Name
zinc finger SWIM-type containing 1
There are no transcripts associated with this gene.

View Full Product Details