Product Details

SNP ID
rs111674328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18729032 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTCTGGATCATGCCGCCCCTGC[A/C]CGGCAGAGATGCACAGTGGGGGCAG
Phenotype
MIM: 176879
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IGSF22 PubMed Links

Gene Details

Gene
IGSF22
Gene Name
immunoglobulin superfamily member 22
There are no transcripts associated with this gene.

Gene
PTPN5
Gene Name
protein tyrosine phosphatase, non-receptor type 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039970.1 2017 Intron NP_001035059.1
NM_001278236.1 2017 Intron NP_001265165.1
NM_001278238.1 2017 Intron NP_001265167.1
NM_001278239.1 2017 Intron NP_001265168.1
NM_006906.1 2017 Intron NP_008837.1
NM_032781.3 2017 Intron NP_116170.3
XM_011520411.2 2017 Silent Mutation CGG,CGT R549R XP_011518713.1
XM_017018434.1 2017 Intron XP_016873923.1
XM_017018435.1 2017 Intron XP_016873924.1
XM_017018436.1 2017 Intron XP_016873925.1
XM_017018437.1 2017 Intron XP_016873926.1
XM_017018438.1 2017 Intron XP_016873927.1
XM_017018439.1 2017 Intron XP_016873928.1
XM_017018440.1 2017 Missense Mutation GGC,TGC G496C XP_016873929.1
XM_017018441.1 2017 Intron XP_016873930.1

View Full Product Details