Product Details

SNP ID
rs34477056
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:38384881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGTAGGTGGCAAGGTCTCTGTA[C/T]TGAAGCCAGTCTGATTCCTTCACTC
Phenotype
MIM: 600418
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMPH PubMed Links

Gene Details

Gene
AMPH
Gene Name
amphiphysin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001635.3 2560 Silent Mutation CAA,CAG Q675Q NP_001626.1
NM_139316.2 2560 Silent Mutation CAA,CAG Q633Q NP_647477.1
XM_006715689.3 2560 Silent Mutation CAA,CAG Q1205Q XP_006715752.1
XM_006715690.3 2560 Silent Mutation CAA,CAG Q799Q XP_006715753.1
XM_006715691.3 2560 Intron XP_006715754.1
XM_011515271.2 2560 Silent Mutation CAA,CAG Q841Q XP_011513573.1
XM_017011995.1 2560 Silent Mutation CAA,CAG Q1247Q XP_016867484.1
XM_017011996.1 2560 Silent Mutation CAA,CAG Q901Q XP_016867485.1
Gene
TRG-AS1
Gene Name
T cell receptor gamma locus antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details