Product Details

SNP ID
rs79924817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:104048170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTTCTGCGATTCCTCCCTCTAC[C/T]GTCCCAGTGGCCTTGAAGCACATTG
Phenotype
MIM: 113811
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COL17A1 PubMed Links

Gene Details

Gene
COL17A1
Gene Name
collagen type XVII alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000494.3 Intron NP_000485.3
Gene
MIR936
Gene Name
microRNA 936
There are no transcripts associated with this gene.

View Full Product Details