Product Details

SNP ID
rs34988585
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:90964295 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCAAATTTTTAACATTGCAGAAC[A/G]TATCTTCGTGCATTTCCAGAATGTT
Phenotype
MIM: 601657
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCER1 PubMed Links

Gene Details

Gene
CCER1
Gene Name
coiled-coil glutamate rich protein 1
There are no transcripts associated with this gene.

Gene
EPYC
Gene Name
epiphycan
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004950.4 739 Missense Mutation NP_004941.2
XM_011538008.1 739 Missense Mutation XP_011536310.1

View Full Product Details