Product Details

SNP ID
rs61737733
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:73383961 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTGCGCCGGGATGTGCGCGTGCT[C/T]GTATGGGGAGTGGTGATAGGAGGGC
Phenotype
MIM: 609729
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDZRN3 PubMed Links

Gene Details

Gene
PDZRN3
Gene Name
PDZ domain containing ring finger 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303139.1 2045 Missense Mutation AAG,GAG K567E NP_001290068.1
NM_001303140.1 2045 Missense Mutation AAG,GAG K526E NP_001290069.1
NM_001303141.1 2045 Missense Mutation AAG,GAG K591E NP_001290070.1
NM_001303142.1 2045 Missense Mutation AAG,GAG K586E NP_001290071.1
NM_015009.2 2045 Missense Mutation AAG,GAG K869E NP_055824.1
XM_017005942.1 2045 Missense Mutation AAG,GAG K840E XP_016861431.1

View Full Product Details