Product Details

SNP ID
rs377310681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137010172 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCGTGAGGACGCGGCGGCCCCGCC[C/T]ACCCAGGGCTCCCGCCCCACCTGTG
Phenotype
MIM: 600047
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCA2 PubMed Links

Gene Details

Gene
ABCA2
Gene Name
ATP binding cassette subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001606.4 Intron NP_001597.2
NM_212533.2 Intron NP_997698.1
XM_006716996.3 Intron XP_006717059.1

View Full Product Details