Product Details

SNP ID
rs151248695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:45758832 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCGTTCTCCCAAGGGAGTTTTC[C/T]TCCTGCACACCTTCCTCATCTTCTT
Phenotype
MIM: 605616
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC6A20 PubMed Links

Gene Details

Gene
SLC6A20
Gene Name
solute carrier family 6 member 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020208.3 2363 UTR 3 NP_064593.1
NM_022405.3 2363 UTR 3 NP_071800.1
XM_011533847.2 2363 UTR 3 XP_011532149.1
XM_011533848.2 2363 Intron XP_011532150.1

View Full Product Details