Product Details
- SNP ID
-
rs61729473
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:81458645 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACAATTTTTCTTTTGATTGTTGTC[T/C]CTTTGCCAAGCCATTTTTGGACGTG
- Phenotype
-
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FABP9
PubMed Links
Gene Details
- Gene
- FABP9
- Gene Name
- fatty acid binding protein 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001080526.1 |
305 |
Missense Mutation |
GAG,GGG |
E102G |
NP_001073995.1 |
View Full Product Details